Learn More
Ruthenium(IV) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
£100.68 - £568.83
Chemical Identifiers
CAS | 12166-20-0 |
---|---|
Molecular Formula | RuS2 |
Molecular Weight (g/mol) | 165.19 |
MDL Number | MFCD00799846 |
InChI Key | XNQLQKWNBPVVGB-UHFFFAOYSA-N |
Synonym | ruthenium iv sulfide, ruthenium sulfide, bis sulfanylidene ruthenium, dithioxoruthenium, ruthenium disulphide, disulfanylideneruthenium |
PubChem CID | 82958 |
SMILES | S=[Ru]=S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384758
|
Thermo Scientific Alfa Aesar
043098.03 |
1 g | N/A | ||||||
11394758
|
Thermo Scientific Alfa Aesar
043098.06 |
5 g |
£568.83
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ruthenium(IV) sulfide is used to prepare sintered material, which is a semiconductor. It acts as an electrode and posses high catalytic activity for oxygen evolution from aqueous solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12166-20-0 | |
165.19 | |
XNQLQKWNBPVVGB-UHFFFAOYSA-N | |
82958 |
RuS2 | |
MFCD00799846 | |
ruthenium iv sulfide, ruthenium sulfide, bis sulfanylidene ruthenium, dithioxoruthenium, ruthenium disulphide, disulfanylideneruthenium | |
S=[Ru]=S |
Specifications
12166-20-0 | |
RuS2 | |
UN3178 | |
XNQLQKWNBPVVGB-UHFFFAOYSA-N | |
disulfanylideneruthenium | |
82958 | |
99.9% | |
Ruthenium (IV) sulfide |
Powder | |
MFCD00799846 | |
ruthenium iv sulfide, ruthenium sulfide, bis sulfanylidene ruthenium, dithioxoruthenium, ruthenium disulphide, disulfanylideneruthenium | |
S=[Ru]=S | |
165.19 | |
165.19 | |
(metals basis) |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.