Learn More
Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver, Silver Nitrate #2 Solution
Determine the presence of spirochetes and nonfilamentous bacteria with Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver.
£50.25 - £270.00
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
10485709
|
Epredia™
87009 |
Steiner-Steiner Silver Kit |
£270.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12543657
|
Epredia™
88033 |
Silver Nitrate Solution #1 | N/A | ||||||
12553657
|
Epredia™
88034 |
Gum Mastic Solution | N/A | ||||||
12510037
|
Epredia™
88035 |
Hydroquinone | N/A | ||||||
12513677
|
Epredia™
88054 |
Sensitizer Solution | N/A | ||||||
Description
- Kit does not contain uranyl nitrate
- Background is stained yellow to brown
- Bacteria, Donovan bodies and fungi all stain black
- Kit formulation and detailed procedures provide consistent and reproducible results