Learn More
Epredia™ Richard-Allan Scientific™ Signature Series Eosin-Y 7111
Create an ideal visual contrast to Hematoxylin with the fast-acting Epredia™ Richard-Allan Scientific™ Signature Series Eosin-Y counterstain.
£38.63
Specifications
Type Eosin Stain
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12677756
View Documents
Epredia™
7111
1 pt. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Bright, vivid cytoplasmic staining
  • Creates three distinct shades of pink
  • Unmatched color contrast between the nuclei and cytoplasm
  • Various shades of red and pink differentiate muscle, red blood cells and connective tissue
  • Strict quality assurance guarantees product consistency and longevity
  • Quantitative testing eliminates need for adjustments in dilution or time
Available in 5 liter packaging
  • Reduces plastic by 90%
  • Reduces stain spills
  • Increases stain stability
  • Saves 35% storage space versus traditional packaging
Specifications
Specifications
Eosin Stain