Learn More
Epredia™ Richard-Allan Scientific™ Cover Glass
Reduces defect costs that range from wasted resources to lost time with a consistent, reliable workflow, which includes quality cover glass.
Brand: Epredia™ 152460
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Ordering Information:
One oz. pack
Specifications
24 mm | |
#1.5 (0.16 to 0.19 mm) | |
Cvr Glass no.1.5 24x60 10 Oz/cs |
60 mm | |
60 x 24 mm |