missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Reversible Microtube Racks with Lid

Store samples securely and rapidly identify them with this reversible microtube rack with lid, which features a low-profile design with an alphanumeric grid.

Brand:  Thermo Scientific™ 8640

 View more versions of this product

Product Code. 10578862

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Store samples securely and rapidly identify them with Thermo Scientific™ Reversible Microtube Racks with Lid. These racks feature a stable, low-profile design with an alphanumeric grid.

  • Hold 0.2mL tubes on one side, 0.5mL tubes on the other
  • Lid protects samples and allows stacking
  • 100% Recycled plastic
  • Several colors available
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Reversible Microtube Rack
Purple
Recycled Plastic
12 Racks
Reversible Microtube Rack
Tubes (0.2 mL), Tubes (0.5 mL)
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.