Learn More
Restek™ Syringe Adapter Kit
For Single-Stage VOC Regulator
£87.12
Specifications
For Use With (Application) | Use to withdraw sample from a high pressure cylinder after pressure reduction through the high purity VOC single stage regulator |
---|---|
Includes | One Nickel-plated Brass 0.25 in. NPT to female luer fitting, which can be used with an A-2 Luer syringe, one Stainless Steel, 0.25 in. NPT x 0.125 in. compression fitting with septum (can be used with any syringe needle) |
Includes: One nickel-plated brass 1/4 in. NPT to female luer fitting, which can be used with an A-2 Luer syringe, One stainless steel, 1/4 in. NPT x 1/8 in. compression fitting with septum (can be used with any syringe needle).
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
13106852
|
Restek™
21118 |
Syringe Adaptor Kit | VOC single stage regulators |
£87.12
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Use to withdraw sample from a high-pressure cylinder after pressure reduction through the high-purity VOC single-stage regulator
Specifications
Use to withdraw sample from a high pressure cylinder after pressure reduction through the high purity VOC single stage regulator | |
One Nickel-plated Brass 0.25 in. NPT to female luer fitting, which can be used with an A-2 Luer syringe, one Stainless Steel, 0.25 in. NPT x 0.125 in. compression fitting with septum (can be used with any syringe needle) |