Learn More
Restek™ Inlet Liners for Splitless Injection—Gooseneck Splitless
Useful for splitless mode of operation and slow injections, small (<2μL) sample volumes, and high-boiling point solvent systems
Brand: Restek™ 20800-202.25
Code : Z3
Additional Details : Weight : 1.00000kg
Descripción
- Compatible with Agilent™/Finnigan columns
- Liners should be packed with wool or fused silica beads to trap nonvolatile residue and prevent column contamination when analyzing dirty samples
Especificaciones
Glass Wool | |
25/Pk. | |
0.4 cm | |
Agilent GCs Equipped with Split/Splitless Inlets |
IP Gooseneck Splitless Single Taper Inlet Liner; For trace samples greater than 2 μL | |
78.5 mm | |
0.65 cm |