Learn More
Restek™ Inlet Liner Removal Tool
Easily remove liner from injector.
Brand:  Restek™ 20181
Code : Z3
Additional Details : Weight : 1.00000kg
Product Code. 13061610
  • £56.87 / Pack of 3
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
  • Made from high-temperature silicone
  • Won’t chip or crack the liner
Specifications
Specifications
Inlet Liner Removal Tool
3/Pk.
Silicone