Learn More
Restek™ Flammable Gas Flash Arrestor - Factory Mutual Approved
Valve cuts off gas supply in the event of a flashback.
Brand: Restek™ 21334
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- Can be reset and reused after flashback has occurred
- Brass body
- 0.25 in. Female NPT
- 316 stainless steel valve spring
- Polyethylene temperature sensor
- Maximum pressure: Hydrogen, 50psig
- Meets safety requirements of BS6158, EN730, ISO 5175, AS 4603 and are UL Listed 23Y5
Specifications
Flammable gases | |
Hydrogen Flashback Arrestor | |
Brass | |
BS6158, EN730, ISO 5175, AS 4603, UL Listed 23Y5 |