Learn More
Trajan Scientific and Medical™ Replacement Needle and Plunger Kit for Shimadzu Autosampler Syringes
Repair kit for Shimadzu Removable Needle Autosampler Syringes.
Brand: Trajan Scientific and Medical™ 033745
Code : 13
Additional Details : Weight : 0.50000kg
Description
For use with Shimadzu Removable Needle Syringes for AOC14, AOC17 and AOC 20 Autosamplers
- Includes spare needle and plunger
- Plunger extends to needle tip
- Sample contained in needle only
- Zero dead volume
- Needle and plunger must be replaced together
Specifications
23 G | |
Replacement Needle Kit | |
Syringes (0.5μL) |
0.63 mm | |
Cone |