Learn More
Restek™ Replacement Chemical Traps for Agilent™ Gas Chromatographs
Easy to install replacement chemical traps attach to the same fittings as original Agilent GC equipment
Brand:  Restek™ 22800
Code : Z3
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13147242
  • £78.65 / Pack of 2
Estimated Shipment: 10-06-2024
to see stock.
Description
Description
  • Built-in frits and adsorbents remove both moisture and hydrocarbons
  • Trap can be regenerated to remove contaminants and restore performance
  • Also available is a Replacement 32 in. Split Line with all installation hardware included
Specifications
Specifications
Replacement 32 in. Split Vent Line
Gas Chromatography
Agilent GCs (replacement for 19251-80525 or G1544-20620)