Learn More
Thermo Scientific™ Remel™ Calibrated Inoculating Loops
Thermo Scientific™ Remel Calibrated Inoculating Loops come in a variety of materials: platinum, twisted nichrome and plastic.Some with color coded with insulated handle.
POA
Description
Thermo Scientific™ Remel Calibrated Inoculating Loops are available in platinum, twisted nichrome and and plastic. Some are color coded with insulated handle.
- Platinum, twisted nichrome or plastic
- With or without insulated handle
- Some are color coded