missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REM2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | REM2 |
---|---|
Concentration | 0.44 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16863955
|
Proteintech
25371-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16853955
|
Proteintech
25371-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Binds GTP saturably and exhibits a low intrinsic rate of GTP hydrolysis.Specifications
REM2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8IYK8 | |
161253 | |
REM2 Fusion Protein Ag21991 | |
Primary | |
-20°C | |
REM2 |
0.44 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GTP binding protein REM 2, REM2 | |
REM2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title