missing translation for 'onlineSavingsMsg'
Learn More

Reichert Instruments Refractometer 230V 50Hz TS400

Brand:  Reichert Instruments 13740000

Refractometer 230V 50Hz TS400

Code : 04

Additional Details : Weight : 1.00000kg

Product Code. 12968584

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Description

Description

Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers