missing translation for 'onlineSavingsMsg'
Learn More

Reelin Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA565442

Product Code. 15988184

  • £453.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: TFCEPYGPREL ITTGLNTTTA SVLQFSIGSG SCRFSYSDPS IIVLYAKNNS ADWIQLEKIR APSNVSTIIH ILYLPEDAKG ENVQFQWKQE NLRVGEVYE Highest antigen sequence identity to the following orthologs - mouse 90%, rat 94%.

Reelin in an extracellular matrix serine protease that plays a role in layering of neurons in the cerebral cortex and cerebellum. Regulates microtubule function in neurons and neuronal migration. Affects migration of sympathetic preganglionic neurons in the spinal cord, where it seems to act as a barrier to neuronal migration. Enzymatic activity is important for the modulation of cell adhesion. Binding to the extracellular domains of lipoprotein receptors VLDLR and LRP8/APOER2 induces tyrosine phosphorylation of DAB1 and modulation of TAU phosphorylation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Reelin
Polyclonal
Unconjugated
RELN
ETL7; extracellular matrix serine protease; LIS2; PRO1598; Reelen; reeler; Reeler protein; Reelin; RELN; Rl
Rabbit
Antigen affinity chromatography
RUO
5649
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P78509
RELN
Recombinant Human Reelin
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only