missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Baculoviral IAP repeat-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) Baculoviral IAP repeat-containing protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7547-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16079310

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001333799.1
Baculoviral IAP repeat-containing protein 1
Research Use Only
NAIP
Recombinant Protein
E. coli
EAKRLKTFVT YEPYSSWIPQ EMAAAGFYFT GVKSGIQCFC CSLILFGAGL TRLPIEDHKR FHPDCGFLLN KDVGNIAKYD IRVKNLKSRL RGGKMRYQEE EARLASFRNW PFYVQGISPC VLSEAGFVFT GKQDTVQCFS CGGCLGNWEE GDDPWKEHAK WFPKCEFLRS KKSSEEITQY IQSYKGFVDI TGEHFVNSWV QRELPMASAY CNDSIFAYEE LRLDSFKDWP RESAVGVAAL AKAGLFYTGI KDIVQCFSCG GCLEKWQEGD DPLDDHTRCF PNCPFL
Greater than 90% as determined by SDS-PAGE.
4671
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers