missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chikungunya Virus CHIKV E1 Protein

A cDNA sequence encoding the CHIKV E1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11405-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15997158

  • £660.00 / 500µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CHIKV E1 Protein
Research Use Only
Recombinant Protein
E. coli
CHIKV E1 protein solution in 1xD-PBS, pH 7.4, 0.1% Thimerosal, 5mM EDTA, 1microg/ml of Leupeptin, and Pepstatin A.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Chikungunya Virus
Untagged
Protein is 95% pure as determined by 12.5% SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers