Learn More
Epredia Alcohol, Reagent; Thermo Scientific Shandon; Filtered, quality reagent consists of: 90%-Absolute ethanol, 5%- Methanol, 5%-Isopropanol; Absolute ethanol replacement; For pathology, staining procedures; No license required by Bureau of ATF
Brand: Epredia 9990520
Alcohol, Reagent; Thermo Scientific Shandon; Filtered, quality reagent consists of: 90%-Absolute ethanol, 5%- Methanol, 5%-Isopropanol; Absolute ethanol replacement; For pathology, staining procedures; No license required by Bureau of ATF
Code : NEW
Additional Details : CAS Number : 64-17-5 Weight : 3.20000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Description