Learn More
RDH10 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14644-1-AP-20UL
This item is not returnable.
View return policy
Description
The RDH10 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RDH10. This antibody recognizes human, mouse, rat antigen. The RDH10 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
RDH10 | |
Polyclonal | |
Unconjugated | |
20 μL | |
157506 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
300 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |