missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGF beta-1 Rat ELISA Kit
For the quantitative detection of rat TGF-beta1.
£451.00 - £2400.00
Specifications
Accession Number | P17246 |
---|---|
Product Type | ELISA Kit |
Sample Type | Plasma, Serum, Supernatant |
Target Species | Rat |
For Use With (Application) | ELISA |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to rat TGF-beta1
Biotin-Conjugate anti-rat TGF-beta1 polyclonal antibody
Streptavidin-HRP
Rat TGF-beta1 Standard lyophilized, 4000pg/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrat Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15512057
|
Invitrogen™
BMS623-3 |
96 Tests |
£451.00
96 tests |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
15542057
|
Invitrogen™
BMS623-3TWO |
2 x 96 Tests |
£580.00
Pack of 2 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15522057
|
Invitrogen™
BMS623-3FIVE |
5 x 96 Tests |
£1444.00
Pack of 5 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15532057
|
Invitrogen™
BMS623-3TEN |
10 x 96 Tests |
£2400.00
Pack of 10 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistencySpecifications
P17246 | |
Plasma, Serum, Supernatant | |
ELISA | |
59086 | |
6.8% | |
RUO | |
2°C to 8°C | |
Rat | |
4 hr. |
ELISA Kit | |
Rat | |
Colorimetric Microplate Reader | |
6.8% | |
HRP | |
Plasma, 20 μL; Serum, 20 μL; Supernatant, 20 μL | |
Camurati-Engelmann disease, DPD1 | |
1 hr. 20 min. |