missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Rat Nampt (Q80Z29) Full-length Recombinant Protein
Used for Func, SDS-PAGE
Brand: Abnova™ P5147.10ug
Additional Details : Weight : 0.00010kg
Specifications
Q80Z29 | |
Lyophilized | |
53.0kDa | |
2 ug/lane in SDS-PAGE | |
Store at -20°C on dry atmosphere. After reconstitution with PBS, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
<0.1ng/μg | |
Nampt | |
Mimics insulin effects in differentiating 3T3-L1 cells. | |
Recombinant | |
Lyophilized |
Functional Study, SDS-PAGE | |
297508 | |
Nampt (Rat) Recombinant Protein | |
10 ug | |
RUO | |
Pbef/Pbef1 | |
Nampt | |
E. coli | |
None | |
≥95% by SDS-PAGE |