Learn More
LPHN2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LPHN2.
Brand:  Abnova PAB25949.50ug
Product Code. 16096127
  • £527.00 / 50µg
Explore more special offers
Description
Description
This gene encodes a member of the latrophilin subfamily of G-protein coupled receptors (GPCR). Latrophilins may function in both cell adhesion and signal transduction. In experiments with nonhuman species, endogenous proteolytic cleavage within a cysteine-rich GPS (G-protein-coupled-receptor proteolysis site) domain resulted in two subunits (a large extracellular N-terminal cell adhesion subunit and a subunit with substantial similarity to the secretin/calcitonin family of GPCRs) being noncovalently bound at the cell membrane. While several transcript variants have been described, the biological validity of only one has been determined. [provided by RefSeq
Specifications
Specifications
LPHN2
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of LPHN2.
In PBS (0.09% sodium azide)
CIRL2/CL2/LEC1/LPHH1
Rabbit
Immunoaffinity chromatography
RUO
23266
Human
Liquid
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (18-45 ug/mL) The optimal working dilution should be determined by the end user.
LPHN2
LPHN2
A synthetic peptide corresponding to 16 amino acids at N-terminus of human LPHN2.
50 μg
Primary
BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.