missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB7A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£132.00 - £349.00
Specifications
Antigen | RAB7A |
---|---|
Concentration | 380 μg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16876935
|
Proteintech
55469-1-AP-20UL |
20 μL |
£132.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16866935
|
Proteintech
55469-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RAB7A antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human RAB7A. This antibody recognizes human, mouse, rat antigen. The RAB7A antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
RAB7A | |
Western Blot, Immunoprecipitation, Immunohistochemistry | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
380 μg/mL | |
Polyclonal | |
Rabbit | |
7879 | |
Primary |