missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Propylene oxide, 99%, Thermo Scientific Chemicals

Product Code. 11444947
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(R)-(+)-Propylene oxide is widely utilized in the manufacture of polyether polyols and propylene glycol, which is used to make the polyurethane plastics. It finds application as a fumigant for the sterilization of packaged foods and plastic medical instruments. Combine with ethanol, it used to make biological samples for electron microscopy. It also used in model aircraft and surface vehicles as a glow fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-Propylene oxide is widely utilized in the manufacture of polyether polyols and propylene glycol, which is used to make the polyurethane plastics. It finds application as a fumigant for the sterilization of packaged foods and plastic medical instruments. Combine with ethanol, it used to make biological samples for electron microscopy. It also used in model aircraft and surface vehicles as a glow fuel.

Solubility
Miscible with water and many organic solvents.

Notes
Highly flammable. Keep away from sources of ignition. The substance may polymerize under the influence of bases, acids and metal chlorides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15448-47-2
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00066211
InChI Key GOOHAUXETOMSMM-GSVOUGTGSA-N
Synonym r-+-propylene oxide, r-propylene oxide, 2r-2-methyloxirane, r-2-methyloxirane, r-methyloxirane, +-propylene oxide, r-epoxypropane, +-methyloxirane, r-+-1,2-epoxypropane, r-1,2-epoxypropane
PubChem CID 146261
ChEBI CHEBI:28985
IUPAC Name (2R)-2-methyloxirane
SMILES CC1CO1

Specifications

Density 0.829
Boiling Point 34°C to 35°C
Flash Point −37°C (−35°F)
Odor Ether-like
Quantity 25 g
UN Number UN1280
Beilstein 79764
Merck Index 14,7856
Solubility Information Miscible with water and many organic solvents.
Formula Weight 58.08
Percent Purity 99%
Chemical Name or Material (R)-(+)-Propylene oxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.