missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human/Mouse/Rat Activin A Protein
CHO derived
Brand: R&D Systems™ 338-AC-050
Additional Details : Weight : 0.00970kg
Description
Measured by its ability to induce hemoglobin expression in K562 human chronic myelogenous leukemia cells. Schwall, R.H. et al. (1991) Method Enzymol. 198:340. The ED50 for this effect is 0.2-1.2 ng/mL.Specifications
P08476 | |
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | |
M.W. (Observed): 14 kDa, reducing conditions 24 kDa, non-reducing conditions; M.W. (theoretical): 13 kDa (monomer) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
<0.01 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
Reconstitute at 100-500 μg/mL in sterile 4 mM HCl. | |
Activin A |
Bioactivity | |
3624 | |
Activin A | |
Chinese Hamster Ovary cell line, CHO-derived Activin A protein Gly311-Ser426 | |
Disulfide-linked homodimer | |
INHBA | |
Recombinant | |
>95%, by SDS-PAGE with silver staining. |