missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-(Dimethylamino)pyrrolidine, 98%, Thermo Scientific Chemicals

£104.00 - £384.00

Chemical Identifiers

CAS 132958-72-6
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00191347
InChI Key AVAWMINJNRAQFS-ZCFIWIBFSA-N
Synonym r-3-dimethylaminopyrrolidine, r-+-3-dimethylamino pyrrolidine, r-n,n-dimethylpyrrolidin-3-amine, 3r-+-3-dimethylamino pyrrolidine, 3r-n,n-dimethylpyrrolidin-3-amine, 3r-3-dimethylaminopyrrolidine, r-3-dimethylamino pyrrolidine, r-dmap, 3r-n,n-dimethyl-3-pyrrolidinamine, r-dimethyl-pyrrolidin-3-yl-amine
PubChem CID 2758521
IUPAC Name (3R)-N,N-dimethylpyrrolidin-3-amine
SMILES CN(C)C1CCNC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10021614
View Documents
Thermo Scientific Acros
426990010
1 g Glass bottle
£104.00
1g
Please to purchase this item. Need a web account? Register with us today!
10694755
View Documents
Thermo Scientific Acros
426990050
5 g Glass bottle
£384.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

132958-72-6
114.19
AVAWMINJNRAQFS-ZCFIWIBFSA-N
2758521
CN(C)C1CCNC1
C6H14N2
MFCD00191347
r-3-dimethylaminopyrrolidine, r-+-3-dimethylamino pyrrolidine, r-n,n-dimethylpyrrolidin-3-amine, 3r-+-3-dimethylamino pyrrolidine, 3r-n,n-dimethylpyrrolidin-3-amine, 3r-3-dimethylaminopyrrolidine, r-3-dimethylamino pyrrolidine, r-dmap, 3r-n,n-dimethyl-3-pyrrolidinamine, r-dimethyl-pyrrolidin-3-yl-amine
(3R)-N,N-dimethylpyrrolidin-3-amine

Specifications

132958-72-6
100.0
51.7°C
C6H14N2
r-3-dimethylaminopyrrolidine, r-+-3-dimethylamino pyrrolidine, r-n,n-dimethylpyrrolidin-3-amine, 3r-+-3-dimethylamino pyrrolidine, 3r-n,n-dimethylpyrrolidin-3-amine, 3r-3-dimethylaminopyrrolidine, r-3-dimethylamino pyrrolidine, r-dmap, 3r-n,n-dimethyl-3-pyrrolidinamine, r-dimethyl-pyrrolidin-3-yl-amine
CN(C)C1CCNC1
114.19
114.19
(R)-(+)-3-(Dimethylamino)pyrrolidine
97.5
166°C
97.5% min. (GC)
MFCD00191347
AVAWMINJNRAQFS-ZCFIWIBFSA-N
(3R)-N,N-dimethylpyrrolidin-3-amine
2758521
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Product Identifier
  • (R)-(+)-3-(Dimethylamino)pyrrolidine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only