missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Plastic Coated Borosilicate Glass Wide Neck Round Media Bottles Without Cap
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
2000 mL
250 mL
500 mL
5000 mL
Description
Manufactured from Borosilicate glass with clear plastic coating to provide added protection against mechanical breakage. Pyrex™ Plastic Coated Borosilicate Glass Wide Neck Round Media Bottles Without Cap reduces the hazard of flying glass and liquid splashes in the event of breakage. Coating withstands steam sterilisation up to 135° C. Bottles are supplied without cap or pouring ring.
- Bottles only. Supplied without cap or pouring ring
- Borosilicate glass with clear plastic coating to provide added protection against mechanical breakage
- Reduces the hazard of flying glass and liquid splashes in the event of breakage
- Printed with retrace code: downloadable batch certificate
- Coating withstands steam sterilisation up to 135°C
Specifications
Specifications
Capacity (Metric) | 1000 mL |
Coating | Plastic |
Color | Clear |
For Use With (Application) | Ideal for mixing and storage of culture medium |
Material | Borosilicate Glass |
Certifications/Compliance | ISO 4796 |
Autoclavable | Autoclavable |
Graduated | Yes |
Mouth | Wide |
Shape | Round |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'