missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Funnel with Interior Ribs

Constructed of Pyrex™ borosilicate glass. Pyrex™ Funnel with Interior Ribs is useful for increasing filtration rate. Suits filter paper of various diameters.

Brand:  Pyrex 2150/03M

 View more versions of this product

Product Code. 12529155

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • With interior 'ribs' to increase the filtration rate
  • Robust construction in Pyrex™ borosilicate glass
  • Stem length of 150mm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

65mm
Pyrex Glass, Borosilicate Glass
(Filter paper) 70 to 90mm
Fluted filter funnel
8mm
196 mm
150mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions