missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Gooch Crucible with Integral Glass Sintered Disc

Ideal for drying precipitates to constant mass at 110°C. Pyrex™ Gooch Crucible includes an integral sintered disc.

Brand:  Pyrex 3660/04M

 View more versions of this product

Product Code. 11377974

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ideal for drying precipitates to constant mass at 110°C. PyrexTM Gooch Crucible includes an integral sintered disc.

  • Ideal for drying precipitates to constant mass at 110°C
  • Also suitable for ignition work up to 450°C (see safety notice)
  • With integral glass sintered disc
  • Manufactured from chemically resistant Pyrex™ borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications

Specifications

With Integral Glass sintered disc
Borosilicate Glass
450°C
53mm
2
30mm
Ideal for drying precipitates to constant mass at 110°C and for ignition work up to 450°C
30 mL
Gooch crucible
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers