missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYREX™ 95mm Diameter Glass Buchner Filter Funnel
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 95mm Glass Buchner Filter Funnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
£510.00 - £2055.00
Specifications
Capacity (Metric) | 500 mL |
---|---|
Diameter (Metric) Stem | 19mm |
Height (Metric) | 250 mm |
Diameter (Metric) Disc | 95mm |
Includes | Integrated filter plate sintered Glass disc |
Product Code | Brand | Porosity | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Pore Size | Price | Quantity & Availability | ||||
11357994
|
Pyrex
3710/01 |
0 | 160 to 250μm |
£510.00
Pack of 5 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||||
12663222
|
Pyrex
3710/04 |
2 | 40 to 100μm |
£515.00
Pack of 5 |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
12673222
|
Pyrex
3710/06 |
3 | 16 to 40μm |
£753.00
Pack of 5 |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
15577710
|
Pyrex
3710/02 |
1 | 100 to 160μm |
£2055.00
Pack of 20 |
Estimated Shipment: 08-11-2024
Log in to see stock. |
||||
15587710
|
Pyrex
3710/08 |
4 | 10 to 16μm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15597710
|
Pyrex
3710/10 |
5 | 1 to 16μm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 95mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
- Comprehensive range of Buchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions - No filter papers required - Highly resistant to most corrosive reagents including ammonia and concentrated sulfuric acid
- Choice of disc diameters and porosities
Specifications
500 mL | |
250 mm | |
Integrated filter plate sintered Glass disc |
19mm | |
95mm |