Learn More
VACUUBRAND™ PVDF/PTFE In-Line Isolation Valve
These straight-through valves with electromagnetic (solenoid) drive are used for operating cycles with short intervals and, therefore, are often used as vacuum control valves
Brand:  VACUUBRAND™ 20674210
Code : A0
Additional Details : Weight : 0.80000kg
Product Code. 10190313
  • £790.00 / Each
Estimated Shipment: 10-07-2024
to see stock.
Add to basket
Description
Description
These straight-through valves with electromagnetic (solenoid) drive are used for operating cycles with short intervals and, therefore, are often used as vacuum control valves. The straightforward design provides a valve with low leakage rate. The valves in version VV C are made of materials with excellent chemical resistance. The valve sealing of the VV-B 6C is made of a special fluoroelastomer with better form stability than common PTfE, much higher chemical resistance than fKM and excellent long-term leak tightness characteristics.
  • version C with excellent chemical resistance
  • high operating cycles
  • significantly better long term tightness even after long operating time
  • easy to clean
  • versatile connection alternatives via small flange or hose nozzle
Specifications
Specifications
PVDF, PTFE
1 Each
In-Line Isolation Valve