Learn More
Buerkle™ PVC Standard Bung
Laboratory standard bung made of PVC with good chemical resistance against acids and alkalis.
Brand:  Buerkle™ 9763-4249
4.85 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17290644
  • £5.65 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Alert:
Attention! Make sure to pay attention to the List of Chemical Resistance!
Description
Description
  • PVC, soft, red
Specifications
Specifications
34 mm
42 mm
49 mm
Red