Learn More
Cole-Parmer™ PTFE Screen Gaskets
Protect your pump, valves and other components in fluid line
£167.00 - £201.00
Product Code | Brand | Diameter (Metric) Outside | Mesh Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Mesh Size | Price | Quantity & Availability | ||||
16629295
|
Cole-Parmer™
3053880 |
1.27 cm | 100 |
£197.00
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
16679275
|
Cole-Parmer™
3053804 |
2.54 cm | 10 |
£167.00
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
16659285
|
Cole-Parmer™
3053844 |
2.54 cm | 4 |
£174.00
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
16689275
|
Cole-Parmer™
3053806 |
3.81 cm | 10 |
£183.00
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
16639295
|
Cole-Parmer™
3053886 |
3.81 cm | 100 |
£183.00
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
16699275
|
Cole-Parmer™
3053808 |
5.08 cm | 10 |
£201.00
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
16639285
|
Cole-Parmer™
3053828 |
5.08 cm | 20 |
£201.00
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
16679285
|
Cole-Parmer™
3053848 |
5.08 cm | 40 |
£201.00
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
16649295
|
Cole-Parmer™
3053888 |
5.08 cm | 100 |
£201.00
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
Description
- Pre-filter before critical membrane filters
- Generate turbulence for enhanced mass or heat transfer
- Purify in chromatography applications
Specifications
PTFE | |
PTFE Gasket |