Learn More
VITLAB™ PTFE Polygonal Magnetic Stirring Bar
Ensure long-lasting durability and prevent demagnetization of Vitlab™ PFTE Polygonal Magnetic Stirring Bars via proper storage and not dropping them on hard surfaces (e.g. steel).
£25.25 - £98.70
Specifications
Material | PTFE |
---|---|
Shape | Polygon |
For Use With (Application) | For preparation, separation and concentration |
Includes | Permanent magnet AlNiCo V core |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
16513602
|
VITLAB™
301097 |
3 mm | 10 mm |
£25.65
Pack of 10 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
10264171
|
VITLAB™
301697 |
6 mm | 10 mm |
£26.50
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
10375661
|
VITLAB™
301597 |
4.5 mm | 12 mm |
£26.50
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
10674602
|
VITLAB™
301797 |
6 mm | 15 mm |
£28.60
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
10375711
|
VITLAB™
301997 |
6 mm | 25 mm |
£31.00
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
10789421
|
VITLAB™
302097 |
6 mm | 30 mm |
£31.35
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
10204271
|
VITLAB™
302197 |
8 mm | 40 mm |
£42.25
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
10224221
|
VITLAB™
301897 |
7 mm | 20 mm |
£29.90
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
16583592
|
VITLAB™
300497 |
2 mm | 5 mm |
£25.50
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
10480343
|
VITLAB™
302397 |
7 mm | 60 mm |
£57.75
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
16533602
|
VITLAB™
302497 |
10 mm | 70 mm |
£90.10
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
16543602
|
VITLAB™
302597 |
10 mm | 80 mm |
£98.70
Pack of 10 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
16503602
|
VITLAB™
300897 |
3 mm | 8 mm |
£25.25
Pack of 10 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
Description
- Permanent magnet AlNiCo V core
- Angled shape gives rise to significant turbulence
- Achieves effective mixing, even at low spin rates
- FTE
Specifications
PTFE | |
Polygon | |
For preparation, separation and concentration | |
Permanent magnet AlNiCo V core |