Learn More
BINDER™ Pt 100 Temperature Sensor
For Use With: KB 720, KB 240, KB 400
Brand: BINDER™ 8012-1079
This item is not returnable.
View return policy
Description
- Flexible
- Additional sensor
- Interior
Specifications
Pt 100 temperature sensor - additional flexible Pt 100, interior, for displaying the temperature on the unit display | |
BINDER Chambers KB 720, KB 240, KB 400 | |
Temperature Sensor |