missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSCA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17171-1-AP-20UL
This item is not returnable.
View return policy
Description
PSCA (Prostate Stem Cell antigen) is a cell surface antigen, which is overexpressed in approximately 40% of primary prostate cancers and in as many as 100% of metastatic ones. PSCA is also overexpressed in a majority of transitional cell and pancreatic carcinomas. Antibody directed against PSCA inhibits tumorigenesis, slows tumor growth, prolongs survival and prevents metastasis in a preclinical zenograft model indicating that PSCA may have utility as a prognostic marker and/or therapeutic target in prostate cancer.Specifications
PSCA | |
Polyclonal | |
Unconjugated | |
PSCA | |
PRO232, prostate stem cell antigen, PSCA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8000 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43653 | |
PSCA | |
PSCA Fusion Protein Ag10710 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |