missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13792-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a transcription factor of the PR-domain protein family. It contains a PR-domain and multiple zinc finger motifs. Transcription factors of the PR-domain family are known to be involved in cell differentiation and tumorigenesis. An elevated expression level of this gene has been observed in PC12 cells treated with nerve growth factor, beta polypeptide (NGF). This gene is located in a chromosomal region that is thought to contain tumor suppressor genes.Specifications
PRDM4 | |
Polyclonal | |
Unconjugated | |
PRDM4 | |
PFM1, PR domain containing 4, PR domain containing protein 4, PRDM4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11108, 170820, 72843 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80V63, Q9QZP2, Q9UKN5 | |
Prdm4 | |
PRDM4 Fusion Protein Ag4580 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |