missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Poxygrid™ Wire Test Tube Racks
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
£36.05 - £52.11
Specifications
Product Type | Test Tube Rack |
---|---|
Autoclavable | Autoclavable |
Diameter (Metric) Opening | 16 mm |
Product Code | Brand | Color | Positions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Positions | Price | Quantity & Availability | ||||
15530022
|
Bel-Art™
F18757-0001 |
Blue | 72 |
£47.05
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
||||
13402509
|
Bel-Art™
F18756-0160 |
Blue | 60 |
£52.00
Pack of 24 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
||||
15570022
|
Bel-Art™
F18759-0002 |
Yellow | 108 | N/A | |||||
15500022
|
Bel-Art™
F18755-0001 |
Blue | 48 |
£46.70
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15590012
|
Bel-Art™
F18755-0000 |
Green | 48 | N/A | |||||
15332367
|
Bel-Art™
F18759-0001 |
Blue | 108 |
£46.20
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
15560022
|
Bel-Art™
F18759-0000 |
Green | 108 |
£46.23
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
||||
12972550
|
Bel-Art™
F18757-0000 |
Green | 72 |
£42.55
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
||||
15580022
|
Bel-Art™
F18759-0003 |
Orange | 108 |
£46.68
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
||||
13422509
|
Bel-Art™
F18760-1150 |
Blue | 150 |
£52.11
Pack of 12 |
Estimated Shipment: 30-01-2025
Log in to see stock. |
||||
Description
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
- Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
- Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
- Orange racks can be used for biohazard warning while yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Stores from 40 to 108 tubes (five different diameters of testtubes)
- Autoclavable
Specifications
Test Tube Rack | |
Autoclavable | |
16 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title