Learn More
3M™ Powerflow™ Battery Pack BAT-25
3M™ Powerflow™ Battery Pack BAT-25 is designed for use with 3M™ Powerflow™ Powered Air Respirator 7900PF.
Brand: 3M™ 7000044297
Code : NEW
Additional Details : Weight : 0.79400kg Transport : UN number : 3480 Chem class : 9
Description
- Rechargeable 5 hour, 8.4V Li-ion battery pack
- Rechargeable battery pack
- For use with 3M™ Powered Air Respirator 7900PF
Specifications
Lithium Ion | |
Battery Pack | |
3M™ Powerflow™ Powered Air Respirator 7900PF | |
8.4 V |