missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, Honeywell™

Product Code. 15625990
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a.,≥98.0% (AT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Quantity 1 kg
Linear Formula KSCN
UN Number NONH for all modes of transport
Beilstein 3594799
Formula Weight 97.18g/mol
Percent Purity ≥98% (AT)
Chemical Name or Material Potassium thiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'