missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Potassium thiocyanate, 99%, for biochemistry
£38.79 - £113.57
Chemical Identifiers
CAS | 333-20-0 |
---|---|
Molecular Formula | CKNS |
Molecular Weight (g/mol) | 97.18 |
InChI Key | ZNNZYHKDIALBAK-UHFFFAOYSA-M |
Synonym | potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt |
PubChem CID | 516872 |
ChEBI | CHEBI:30951 |
IUPAC Name | potassium;thiocyanate |
SMILES | C(#N)[S-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12625137
|
Thermo Scientific Chemicals
448002500 |
250 g | Plastic bottle |
£38.79
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12615137
|
Thermo Scientific Chemicals
448000010 |
1 kg | Plastic bottle |
£113.57
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
333-20-0 | |
97.18 | |
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt | |
CHEBI:30951 | |
C(#N)[S-].[K+] |
CKNS | |
ZNNZYHKDIALBAK-UHFFFAOYSA-M | |
516872 | |
potassium;thiocyanate |
Specifications
170.0°C to 179.0°C | |
98.5% min. (Argentometry) | |
KSCN | |
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt | |
ZNNZYHKDIALBAK-UHFFFAOYSA-M | |
potassium;thiocyanate | |
516872 | |
97.18 | |
Biochemical | |
1.8860g/mL |
333-20-0 | |
CKNS | |
15,7807 | |
Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone | |
C(#N)[S-].[K+] | |
97.18 | |
CHEBI:30951 | |
99% | |
1.886 | |
Potassium thiocyanate |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 206-370-1
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Potassium thiocyanate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P233-Keep container tightly closed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : Potassium thiocyanate
RUO – Research Use Only