Learn More
Potassium Pyroantimonate EU Pharmacopoeia Grade Product Code.: 12900524
Potassium Pyroantimonate EU Pharmacopoeia Grade
BIOPHARMA
Product Code. 12900524
125 Pk.
Click to view available options
Quantity:
125 Pk.
Product Code. 12900524
Brand: Fisher Chemical RP 183 125
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Quantity 125 Pk.
compliance-icons
Product Identifier
  • Potassium pyroantimonate Solution European Pharmacopoeia Ref: 1071301 (RP 183)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Product Content Correction
Product Title
Potassium Pyroantimonate EU Pharmacopoeia Grade >