Learn More
Potassium phosphate tribasic, 90+%, pure, Thermo Scientific Chemicals
£33.67 - £222.00
Chemical Identifiers
CAS | 7778-53-2 |
---|---|
Molecular Formula | K3O4P |
Molecular Weight (g/mol) | 212.27 |
InChI Key | LWIHDJKSTIGBAC-UHFFFAOYSA-K |
IUPAC Name | tripotassium phosphate |
SMILES | [K+].[K+].[K+].[O-]P([O-])([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17211312
|
Thermo Scientific Acros
467330010 |
1 kg |
£33.67
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17221312
|
Thermo Scientific Acros
467330050 |
5 kg |
£89.24
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17231312
|
Thermo Scientific Acros
467330250 |
25 kg | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7778-53-2 | |
212.27 | |
tripotassium phosphate |
K3O4P | |
LWIHDJKSTIGBAC-UHFFFAOYSA-K | |
[K+].[K+].[K+].[O-]P([O-])([O-])=O |
Specifications
1340.0°C | |
90+% | |
K3PO4 | |
LWIHDJKSTIGBAC-UHFFFAOYSA-K | |
tripotassium phosphate | |
212.27 | |
Potassium phosphate tribasic |
7778-53-2 | |
K3O4P | |
Solubility aq. soln.: 508 g/L (25°C). Other solubilities: insoluble in alcohol | |
[K+].[K+].[K+].[O-]P([O-])([O-])=O | |
212.27 | |
Pure |
Safety and Handling
GHS H Statement Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-907-1
- Potassium phosphate tribasic
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Phosphoric acid, tripotassium salt
RUO – Research Use Only