missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium oxalate monohydrate, Honeywell™
Purum p.a.,≥99.0% (RT)
£44.89 - £127.45
Chemical Identifiers
CAS | 6487-48-5 |
---|---|
Molecular Formula | C2H2K2O5 |
Molecular Weight (g/mol) | 184.23 |
MDL Number | MFCD00150033 |
InChI Key | QCPTVXCMROGZOL-UHFFFAOYSA-L |
Synonym | potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate |
PubChem CID | 2724193 |
IUPAC Name | dipotassium;oxalate;hydrate |
SMILES | O.[K+].[K+].[O-]C(=O)C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15655970
|
Honeywell Research Chemicals
60425-250G |
250 g |
£44.89
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15645970
|
Honeywell Research Chemicals
60425-1KG |
1 kg |
£127.45
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
6487-48-5 | |
184.23 | |
QCPTVXCMROGZOL-UHFFFAOYSA-L | |
2724193 | |
O.[K+].[K+].[O-]C(=O)C([O-])=O |
C2H2K2O5 | |
MFCD00150033 | |
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate | |
dipotassium;oxalate;hydrate |
Specifications
6487-48-5 | |
(COOK)2 · H2O | |
NONH for all modes of transport | |
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate | |
O.[K+].[K+].[O-]C(=O)C([O-])=O | |
184.23 | |
184.23g/mol | |
Potassium oxalate monohydrate |
C2H2K2O5 | |
MFCD00150033 | |
3752576 | |
QCPTVXCMROGZOL-UHFFFAOYSA-L | |
dipotassium;oxalate;hydrate | |
2724193 | |
≥99% (RT) |
Safety and Handling
P280-P301 + P312 + P330
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Potassium oxalate monohydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Potassium oxalate-1-hydrate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title