missing translation for 'onlineSavingsMsg'
Learn More

Potassium orthovanadate, 99.9% (metals basis), Thermo Scientific Chemicals

£137.69 - £483.41

Chemical Identifiers

CAS 14293-78-8
Molecular Formula K3O4V
Molecular Weight (g/mol) 232.232
MDL Number MFCD00210642
InChI Key FBRINGOENSVSPU-UHFFFAOYSA-N
Synonym Potassium vanadate (ortho)
PubChem CID 22023817
IUPAC Name tripotassium;trioxido(oxo)vanadium
SMILES [O-][V](=O)([O-])[O-].[K+].[K+].[K+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11328308
SDS View Documents
Thermo Scientific Alfa Aesar
039502.14
25 g
£137.69
25g
Please to purchase this item. Need a web account? Register with us today!
11318308
SDS View Documents
Thermo Scientific Alfa Aesar
039502.22
100 g
£483.41
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is employed as in chemical fertilizer industries and used as corrosion inhibitor. It is also used for ammonia synthesis. Used as a catalyst in oil & gas industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H315-H319-H335
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H332-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.

missing translation for 'einecsNumber' : 238-220-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium orthovanadate, 99.9% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.