missing translation for 'onlineSavingsMsg'
Learn More
Potassium nitrate, Puriss., meets analytical spec. of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%, Honeywell Fluka™ Product Code.: 15691170

Potassium nitrate, Puriss., meets analytical spec. of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%, Honeywell Fluka™
BIOPHARMA

Product Code. 15691170 Shop All Honeywell Fluka Products
1 kg
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
This item is not returnable. View return policy

Product Code. 15691170

Brand: Honeywell-Fluka 126481KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitrasShow MoreShow Less
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium;nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Quantity 1 kg
UN Number 1486
Formula Weight 101.1g/mol
Percent Purity 99 to 100.5%
Grade puriss.
Chemical Name or Material Potassium Nitrate, Meets analytical specification of Ph. Eur.BP, USP, FCC

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberL295A Certificate TypeCertificate of Analysis Date29/03/2023 Catalog Number12648-2.5KG,12648-25KG,12648-6X1KG,12648-1KG
Lot NumberA0394708 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number126480025,12648-1KG,12648-25KG,126480010,12648-6X1KG,12648-2.5KG,126480250,126485000,12648010
Lot NumberA0380899 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number126480025,12648-1KG,12648-25KG,126480010,12648-6X1KG,12648-2.5KG,126480250,126485000,12648010
Lot NumberA0436967 Certificate TypeCertificate of Analysis Date30/05/2022 Catalog Number126480025,12648-1KG,12648-25KG,126480010,12648-6X1KG,12648-2.5KG,126480250,126485000,12648010
Lot NumberA0437237 Certificate TypeCertificate of Analysis Date14/12/2021 Catalog Number126480025,12648-1KG,12648-25KG,126480010,12648-6X1KG,12648-2.5KG,126480250,126485000,12648010
compliance-icons
Product Identifier
  • Potassium nitrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Potassium nitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium nitrate, Puriss., meets analytical spec. of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.