Learn More
Potassium methoxide, 90+%, Thermo Scientific Chemicals
£43.32 - £134.67
Chemical Identifiers
CAS | 865-33-8 |
---|---|
Molecular Formula | CH3KO |
Molecular Weight (g/mol) | 70.132 |
MDL Number | MFCD00012416 |
InChI Key | BDAWXSQJJCIFIK-UHFFFAOYSA-N |
Synonym | potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok |
PubChem CID | 23664618 |
IUPAC Name | potassium;methanolate |
SMILES | C[O-].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11347847
|
Thermo Scientific Alfa Aesar
014261.18 |
50 g |
£43.32
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337847
|
Thermo Scientific Alfa Aesar
014261.30 |
250 g |
£134.67
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium methoxide is used as a chemical intermediate. It is also used in biodiesel production and in fuel additives. It acts as a catalyst for the preparation of methyl formate and an alkaline catalyst for dimethyl formamide. Further, it is used as versatile reagents in chemical reactions such as condensation, esterification, alkoxylation and etherification, Claisen condensation, Wolf-Kishner reduction and Stobbe reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
865-33-8 | |
70.132 | |
BDAWXSQJJCIFIK-UHFFFAOYSA-N | |
23664618 | |
C[O-].[K+] |
CH3KO | |
MFCD00012416 | |
potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok | |
potassium;methanolate |
Specifications
865-33-8 | |
>90% | |
MFCD00012416 | |
Soluble in methanol and ethanol. | |
C[O-].[K+] | |
70.132 | |
70.14 | |
Potassium methoxide |
Powder | |
CH3KO | |
potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok | |
BDAWXSQJJCIFIK-UHFFFAOYSA-N | |
potassium;methanolate | |
23664618 | |
Air and moisture sensitive |
Safety and Handling
P234-P235+P410-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H302-H314-H335-X
missing translation for 'dotInformation' : Hazard Class: 4.2; Packaging Group: II
missing translation for 'einecsNumber' : 212-736-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Potassium methoxide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Self-heating substance Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H251-Self-heating: may catch fire.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P235-Keep cool.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P420-Store away from other materials.
Supplemental information
- MIXTURE LIST-Contains : Methanol, potassiumsalt
RUO – Research Use Only