Learn More
Potassium iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
£61.20 - £162.81
Chemical Identifiers
CAS | 7681-11-0 |
---|---|
Molecular Formula | IK |
Molecular Weight (g/mol) | 166.003 |
MDL Number | MFCD00011405 |
InChI Key | NLKNQRATVPKPDG-UHFFFAOYSA-M |
Synonym | potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide |
PubChem CID | 4875 |
ChEBI | CHEBI:8346 |
IUPAC Name | potassium;iodide |
SMILES | [K+].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11326938
|
Thermo Scientific Alfa Aesar
013676.06 |
5 g |
£61.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11316938
|
Thermo Scientific Alfa Aesar
013676.14 |
25 g |
£162.81
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium iodide is a simple iodide salt utilized as a fluorescence quenching agent. It is used in photography and as an analytical reagent, It is used in treating overactive thyroid and protecting the thyroid gland from the effects of radiation from inhaled or swallowed radioactive iodine. It is used as a nutrition supplement to prevent goitre and other iodine deficiency in human body.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7681-11-0 | |
166.003 | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
4875 | |
potassium;iodide |
IK | |
MFCD00011405 | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
CHEBI:8346 | |
[K+].[I-] |
Specifications
1330°C | |
7681-11-0 | |
Beads | |
IK | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
potassium;iodide | |
4875 | |
166.01 | |
1.677 | |
White beads | |
3.12 g/cm3 |
681°C | |
White | |
99.998% (metals basis) | |
MFCD00011405 | |
Soluble in water (148g/100g at 25°C),alcohol (45.5mg/ml),methanol (125mg/ml),acetone (13.1mg/ml at 25°C),and ethanol (19.6mg/ml). | |
[K+].[I-] | |
166.003 | |
CHEBI:8346 | |
Odorless | |
Hygroscopic | |
1.33 hPa (1mm Hg) at 745°C | |
Potassium iodide |
Safety and Handling
P260-P264b-P270-P314-P501c
H372
missing translation for 'einecsNumber' : 213-659-4
missing translation for 'rtecsNumber' : TT2975000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only