missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
12 kg
4 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 4 x 2.5 kg |
Linear Formula | KI |
UN Number | NONH for all modes of transport |
Formula Weight | 166g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Vapor Pressure | 1mmHg (745°C) |
Chemical Name or Material | Potassium Iodide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction