missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution 1M (1N), (Alcoholic), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Chemical Identifiers

MDL Number 3553
View More Specs

Description

Description

Specifications

Chemical Identifiers

3553

Specifications

Colorless to Yellow
3553
HDPE plastic bottle
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium hydroxide solution, 1M in alcohol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
  • EUH071-Corrosive to the respiratory tract.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS
Documents

Documents

Product Certifications